Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01189020_mH
          See other KLRB1 GE Assays ›
          Gene Symbol:
          Klrb1
          Gene Name:
          killer cell lectin-like receptor subfamily B member 1
          Gene Aliases:
          4930431A04Rik, Gm4696, Klrb1g, Klrb6, Ly-55, Ly55, NKR-P1G, Nkrp-1e, Nkrp1g
          Chromosome Location:
          Chr.6: 128706093 - 128764561 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01189020_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klrb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099918.1 NP_001093388.1 5 - 6 528 83
          GenBank mRNA DQ113419.1 - 5 - 6 528 83
          HQ713436.1 - 5 - 6 528 83
          JF429832.1 - 5 - 6 541 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003213.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Process(es)

          receptor activity
          protein binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968890 AK015258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077956 DQ113419.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077956 NM_001099918.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58661 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 AK004652.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 AK082323.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 AK082761.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 AK141692.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 AK145969.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 BC034517.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 NM_007949.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584670 U97572.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 9817 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077956 DQ113419.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077956 NM_001099918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968890 AK015258.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24840465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05769004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05784915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05784914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05873186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05215895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05784902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05486136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05201513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05222418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05201512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05156159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05333441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05873183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05545534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05784900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05873184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24840416 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548302 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548300 - -
          Affymetrix Mouse Clariom S Array PSR0600025119.mm.1 NM_001099918.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025119.mm.1 NM_001099918.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025119.mm.1 TC0600003213.mm.1 MTA TC
          Murine Genome U74v2 Set 170600_PM_at - -
          Murine Genome U74v2 Set 170600_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline