Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01191483_m1
          See other DBNL GE Assays ›
          Gene Symbol:
          Dbnl
          Gene Name:
          drebrin-like
          Gene Aliases:
          Abp1, SH3P7, mAbp1
          Chromosome Location:
          Chr.11: 5788483 - 5800980 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01191483_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbnl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146308.1 NP_001139780.1 11 - 12 1145 121
          NM_001146309.1 NP_001139781.1 11 - 12 1133 121
          NM_013810.3 NP_038838.1 11 - 12 1136 121
          GenBank mRNA AK046073.1 - 11 - 12 1113 121
          AK053796.1 - 11 - 12 1129 121
          AK078082.1 - 11 - 12 1100 121
          AK134136.1 - 11 - 12 1100 121
          AK134487.1 - 11 - 12 1112 121
          AK140752.1 - 11 - 12 1103 121
          AK142818.1 - 11 - 12 1083 121
          AK146920.1 - 11 - 12 1100 121
          AK151096.1 - 11 - 12 1113 121
          AK152110.1 - 11 - 12 1101 121
          AK152204.1 - 11 - 12 1113 121
          AK153072.1 - 11 - 12 1112 121
          AK153389.1 - 11 - 12 1090 121
          AK159984.1 - 11 - 12 1110 121
          AK161973.1 - 10 - 11 1407 121
          AK167956.1 - 11 - 12 1072 121
          AK168898.1 - 11 - 12 1110 121
          AK172471.1 - 11 - 12 1090 121
          AK181313.1 - 2 - 3 121
          AK210402.1 - 2 - 3 121
          AY098595.1 - 11 - 12 1068 121
          BC046430.1 - 11 - 12 1098 4986658 121
          U58884.1 - 11 - 12 1089 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000078.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbnl

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          transport
          endocytosis
          receptor-mediated endocytosis
          synapse assembly
          Rac protein signal transduction
          neuron projection morphogenesis
          actin filament severing
          barbed-end actin filament capping
          actin filament bundle assembly
          podosome assembly
          ruffle assembly
          actin binding
          protein binding
          protein C-terminus binding
          protein domain specific binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_001146308.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_001146308.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_001146309.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_001146309.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_013810.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 NM_013810.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000875.mm.1 TC1100000078.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline