Search
Search
| Gene Symbol: Ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113408.1 | NP_001106879.1 | 4 - 5 | 599 | 94 | |
| NM_010697.1 | NP_034827.1 | 4 - 5 | 784 | 94 | ||
| GenBank mRNA | AB221569.1 | - | 3 - 4 | 141 | 94 | |
| AB250383.1 | - | 3 - 4 | 138 | 94 | ||
| AF030333.1 | - | 4 - 5 | 302 | 94 | ||
| BC013624.1 | - | 4 - 5 | 424 | 3708169 | 94 | |
| CK790183.1 | - | 4 - 5 | 604 | 94 | ||
| U69270.1 | - | 4 - 5 | 784 | 94 | ||
| U70375.1 | - | 3 - 4 | 134 | 94 | ||
| U89488.1 | - | 4 - 5 | 382 | 94 | ||
Set Membership: |
|