Search
Search

Gtf2i| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 13 - 14 | 1434 | 56 | |
| NM_001080747.2 | NP_001074216.1 | 12 - 13 | 1371 | 56 | ||
| NM_001080748.2 | NP_001074217.1 | 11 - 12 | 1314 | 56 | ||
| NM_001080749.2 | NP_001074218.1 | 11 - 12 | 1314 | 56 | ||
| NM_010365.4 | NP_034495.2 | 12 - 13 | 1377 | 56 | ||
| GenBank mRNA | AF017085.1 | - | 13 - 14 | 1078 | 56 | |
| AF043219.1 | - | 11 - 12 | 998 | 56 | ||
| AF043220.1 | - | 10 - 11 | 935 | 56 | ||
| AK145698.1 | - | 3 - 4 | 263 | 56 | ||
| AK147201.1 | - | 11 - 12 | 1272 | 56 | ||
| AY030290.1 | - | 12 - 13 | 1064 | 56 | ||
| AY030291.1 | - | 12 - 13 | 1070 | 56 | ||
| AY030292.1 | - | 11 - 12 | 1007 | 56 | ||
| AY030293.1 | - | 13 - 14 | 1127 | 56 | ||
| BC047387.1 | - | 11 - 12 | 962 | 56 | ||
| BC053044.1 | - | 13 - 14 | 1403 | 5704935 | 56 | |
Set Membership: |
|