Search
Search

Itgb2l| Gene Symbol: Itgb2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008405.3 | NP_032431.2 | 10 - 11 | 1219 | 65 | |
| GenBank mRNA | AF051367.1 | - | 10 - 11 | 1204 | 65 | |
| AK036640.1 | - | 10 - 11 | 1219 | 65 | ||
| AK137534.1 | - | 10 - 11 | 1220 | 65 | ||
| AK158108.1 | - | 10 - 11 | 1220 | 65 | ||
| BC113142.1 | - | 10 - 11 | 1188 | 40047633 | 65 | |
Set Membership: |
|