Search
Search

Nkain4| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141933.1 | NP_001135405.1 | 2 - 3 | 268 | 83 | |
| NM_021426.4 | NP_067401.3 | 2 - 3 | 268 | 83 | ||
| GenBank mRNA | AB030182.1 | - | 2 - 3 | 246 | 83 | |
| AK005262.1 | - | 2 - 3 | 239 | 83 | ||
| AK007632.1 | - | 2 - 3 | 233 | 83 | ||
| AK021096.1 | - | 2 - 3 | 263 | 83 | ||
| BC061107.1 | - | 2 - 3 | 253 | 30295742 | 83 | |
| BC089330.1 | - | 2 - 3 | 220 | 30358178 | 83 | |
| CK625327.1 | - | 2 - 3 | 268 | 83 | ||
| EF058055.1 | - | 2 - 3 | 193 | 83 | ||
| EF058056.1 | - | 2 - 3 | 193 | 83 | ||
| EF058057.1 | - | 2 - 3 | 193 | 83 | ||
Set Membership: |
|