Search
Search
| Gene Symbol: Unk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286006.1 | NP_001272935.1 | 11 - 12 | 1585 | 63 | |
| NM_172569.4 | NP_766157.1 | 11 - 12 | 1624 | 63 | ||
| GenBank mRNA | AK046395.1 | - | 11 - 12 | 1619 | 63 | |
| AK077635.1 | - | 11 - 12 | 1626 | 63 | ||
| AK078095.1 | - | 11 - 12 | 1576 | 63 | ||
| AK129438.1 | - | 11 - 12 | 1602 | 63 | ||
| AK134361.1 | - | 11 - 12 | 1705 | 63 | ||
| BC003195.1 | - | 8 - 9 | 1070 | 63 | ||
| BC039964.1 | - | 11 - 12 | 1586 | 63 | ||
| BC054452.1 | - | 11 - 12 | 1561 | 6491704 | 63 | |
Set Membership: |
|