Search
Search

Exoc3l| Gene Symbol: Exoc3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177788.4 | NP_808456.1 | 7 - 8 | 1524 | 88 | |
| GenBank mRNA | AK050100.1 | - | 7 - 8 | 1525 | 88 | |
| AK140526.1 | - | 3 - 4 | 616 | 88 | ||
| AK172618.1 | - | 6 - 7 | 1191 | 88 | ||
| BC094238.1 | - | 7 - 8 | 1478 | 30604935 | 88 | |
Set Membership: |
|