Search
Search

Neto2| Gene Symbol: Neto2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081324.1 | NP_001074793.1 | 3 - 4 | 373 | 80 | |
| GenBank mRNA | AK017458.1 | - | 3 - 4 | 448 | 80 | |
| AK081462.1 | - | 3 - 4 | 451 | 80 | ||
| AK083512.1 | - | 3 - 4 | 482 | 80 | ||
| AY138991.1 | - | 3 - 4 | 337 | 80 | ||
| BC068168.1 | - | 3 - 4 | 374 | 80 | ||
| BC151050.1 | - | 3 - 4 | 312 | 9087963 | 80 | |
| BC157941.1 | - | 3 - 4 | 312 | 9088121 | 80 | |
| BC172104.1 | - | 3 - 4 | 312 | 9054390 | 80 | |
| BC172139.1 | - | 3 - 4 | 312 | 9054809 | 80 | |
Set Membership: |
|