Search
Search
| Gene Symbol: Wwox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019573.3 | NP_062519.2 | 2 - 3 | 292 | 60 | |
| GenBank mRNA | AF187014.1 | - | 2 - 3 | 243 | 60 | |
| AK018507.1 | - | 2 - 3 | 222 | 60 | ||
| AK019911.1 | - | 2 - 3 | 250 | 60 | ||
| AK046903.1 | - | 2 - 3 | 242 | 60 | ||
| AK050541.1 | - | 2 - 3 | 243 | 60 | ||
| AK078528.1 | - | 2 - 3 | 245 | 60 | ||
| AK083096.1 | - | 2 - 3 | 224 | 60 | ||
| BC014716.1 | - | 2 - 3 | 248 | 4241066 | 60 | |
| CJ058964.1 | - | 2 - 3 | 293 | 60 | ||
Set Membership: |
|