Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029580.3 | NP_083856.1 | 3 - 4 | 793 | 99 | |
GenBank mRNA | AK011576.1 | - | 3 - 4 | 794 | 99 | |
AK049127.1 | - | 3 - 4 | 792 | 99 | ||
AK220389.1 | - | 2 - 3 | 540 | 99 | ||
BC012521.1 | - | 3 - 4 | 794 | 2648293 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |