Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01262007_m1
          See other HSBP1L1 GE Assays ›
          Gene Symbol:
          Hsbp1l1
          Gene Name:
          heat shock factor binding protein 1-like 1
          Gene Aliases:
          1810005K13Rik
          Chromosome Location:
          Chr.18: 80229571 - 80250103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01262007_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsbp1l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136181.1 NP_001129653.1 4 - 5 446 62
          GenBank mRNA AK007343.1 - 3 - 4 292 62
          AK019003.1 - 3 - 4 300 62
          AK141935.1 - 4 - 5 447 62
          BC046504.1 - 3 - 4 318 62
          BC151118.1 - 3 - 4 293 9088031 62
          BC151120.1 - 3 - 4 293 9088033 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001655.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 BC151118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 BC151120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 NM_001136181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 AK007343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 AK019003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 BC151118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 BC151120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 NM_001136181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 AK007343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P18765 AK019003.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122300 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123631 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 BC151118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 BC151120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 NM_001136181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 AK007343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 AK019003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 BC151118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 BC151120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 NM_001136181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 AK007343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P18765 AK019003.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123631 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09065694 - -
          Whole Mouse Genome G4122A A_52_P18765 BC151118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 BC151120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 NM_001136181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 AK007343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 AK019003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 BC151118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 BC151120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 NM_001136181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 AK007343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P18765 AK019003.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127054 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14017094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14076597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13597419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14076587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13893309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13713031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13674719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31863151 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444790_PM_at - -
          Mouse Genome 430 2.0 Array 1444790_PM_at - -
          Mouse Genome 430 2.0 Array 1444790_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66255_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66255_at - -
          Affymetrix Mouse Clariom S Array PSR1800011852.mm.1 NM_001136181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011852.mm.1 NM_001136181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011852.mm.1 TC1800001655.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011853.mm.1 NM_001136181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011853.mm.1 NM_001136181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011853.mm.1 TC1800001655.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011854.mm.1 NM_001136181.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011854.mm.1 NM_001136181.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011854.mm.1 TC1800001655.mm.1 MTA TC
          Mouse Expression Set 430 1444790_at - -
          Mouse Expression Set 430 1444790_PM_at - -
          Murine Genome U74v2 Set 134434_PM_at - -
          Murine Genome U74v2 Set 134434_at - -
          Affymetrix HT Mouse Genome 430B Array 1444790_at - -
          Affymetrix HT Mouse Genome 430B Array 1444790_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926231 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline