Search
Search

Eif3a| Gene Symbol: Eif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010123.3 | NP_034253.3 | 4 - 5 | 717 | 87 | |
| GenBank mRNA | AK082006.1 | - | 4 - 5 | 683 | 87 | |
| AK145132.1 | - | 4 - 5 | 725 | 87 | ||
| AK145629.1 | - | 4 - 5 | 682 | 87 | ||
| AK149482.1 | - | 4 - 5 | 670 | 87 | ||
| AK161634.1 | - | 4 - 5 | 674 | 87 | ||
| AK167010.1 | - | 4 - 5 | 684 | 87 | ||
| AK172899.1 | - | 4 - 5 | 654 | 87 | ||
| U14172.1 | - | 4 - 5 | 667 | 87 | ||
Set Membership: |
|