Search
Search

Trp53bp1| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 6 - 7 | 775 | 64 | |
| NM_013735.4 | NP_038763.3 | 6 - 7 | 775 | 64 | ||
| GenBank mRNA | AJ414734.1 | - | 5 - 6 | 636 | 64 | |
| AK045851.1 | - | 6 - 7 | 690 | 64 | ||
| AK148123.1 | - | 6 - 7 | 735 | 64 | ||
| BC079906.1 | - | 6 - 7 | 709 | 30639375 | 64 | |
Set Membership: |
|