Search
Search

Rnf39| Gene Symbol: Rnf39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099632.1 | NP_001093102.1 | 2 - 3 | 420 | 59 | |
| GenBank mRNA | BC038661.1 | - | 2 - 3 | 490 | 59 | |
| BC051578.1 | - | 2 - 3 | 499 | 59 | ||
| BC132405.1 | - | 2 - 3 | 420 | 40130682 | 59 | |
| BC145677.1 | - | 2 - 3 | 420 | 40130980 | 59 | |
Set Membership: |
|