Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279316_g1
          See other PRDM9 GE Assays ›
          Gene Symbol:
          Prdm9
          Gene Name:
          PR domain containing 9
          Gene Aliases:
          BC012016, Dsbc1, G1-419-29, Meisetz, Rcr1, repro7
          Chromosome Location:
          Chr.17: 15543072 - 15564379 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279316_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdm9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144809.2 NP_659058.2 7 - 8 914 70
          GenBank mRNA AB844106.1 - 7 - 8 914 70
          AB844107.1 - 7 - 8 914 70
          AB844108.1 - 7 - 8 914 70
          AB844109.1 - 7 - 8 920 70
          AB844110.1 - 7 - 8 914 70
          AB844111.1 - 7 - 8 892 70
          AB844112.1 - 7 - 8 892 70
          AB844113.1 - 7 - 8 914 70
          AB844114.1 - 7 - 8 914 70
          AB844116.1 - 917 70
          BC012016.1 - 9 - 10 1055 70
          BC023014.1 - 2 - 3 576 5365459 70
          BC049903.1 - 9 - 10 1055 4501059 70
          FJ212287.1 - 7 - 8 892 70
          GU216230.1 - 7 - 8 892 70
          HQ704390.1 - 7 - 8 892 70
          HQ704391.1 - 7 - 8 892 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001501.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdm9

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic gene conversion
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatogenesis
          positive regulation of reciprocal meiotic recombination
          chromatin modification
          histone methylation
          methylation
          histone lysine methylation
          positive regulation of transcription from RNA polymerase II promoter
          meiotic cell cycle
          histone H3-K4 methylation
          positive regulation of meiosis I
          nucleic acid binding
          methyltransferase activity
          recombination hotspot binding
          transferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700013453.mm.1 NM_144809.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013453.mm.1 NM_144809.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013453.mm.1 TC1700001501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013456.mm.1 TC1700001501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013452.mm.1 NM_144809.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013452.mm.1 NM_144809.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013452.mm.1 TC1700001501.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline