Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01279964_g1
          See other LAMTOR1 GE Assays ›
          Gene Symbol:
          Lamtor1
          Gene Name:
          late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
          Gene Aliases:
          2400001E08Rik, Pdro, p18
          Chromosome Location:
          Chr.7: 101905837 - 101911903 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01279964_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamtor1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025605.3 NP_079881.2 2 - 3 345 76
          GenBank mRNA AK003509.1 - 2 - 3 237 76
          AK009320.1 - 2 - 3 234 76
          AK010247.1 - 2 - 3 278 76
          AK013389.1 - 2 - 3 255 76
          AK019947.1 - 2 - 3 196 76
          AK144476.1 - 2 - 3 244 76
          BC020142.1 - 2 - 3 251 3709051 76
          BC092062.1 - 2 - 3 237 4024506 76
          BC096412.1 - 2 - 3 217 5255882 76
          BC126973.1 - 2 - 3 252 40100355 76

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lamtor1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor recycling
          endosome organization
          lysosome organization
          regulation of cholesterol esterification
          regulation of cholesterol efflux
          cell growth
          positive regulation of TOR signaling
          lysosome localization
          endosome localization
          cellular protein localization
          cholesterol homeostasis
          positive regulation of MAPK cascade
          positive regulation of GTPase activity
          regulation of cholesterol import
          cellular response to amino acid stimulus
          guanyl-nucleotide exchange factor activity
          protein binding
          protein complex scaffold

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376020 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376021 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052185 BC096412.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052185 AK019947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052185 AK080275.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052185 AK144476.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052185 BC020142.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408902 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161809 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052185 BC096412.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052185 AK019947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052185 AK080275.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052185 AK144476.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052185 BC020142.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25376016 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161809 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01591894 - -
          Whole Mouse Genome G4122A A_52_P282785 AK080275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338674 BC096412.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338674 AK144476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338674 BC020142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338674 BC092062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338674 NM_025605.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110929 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102907 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103431 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161809 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05891012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06285643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05890996 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376021 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25376020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06382299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05891012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06320222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06410389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06129837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06221572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06478145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06185149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06181557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06315254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06181554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06315249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06549674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06478141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06272992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376109 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555587 - -
          Affymetrix HT MG-430 PM Array Plate 1434124_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1448718_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555586 - -
          Affymetrix Mouse Gene 1.1 ST Array 10460765 - -
          Mouse Genome 430 2.0 Array 1417463_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448718_PM_at - -
          Mouse Genome 430 2.0 Array 1448718_at - -
          Mouse Genome 430 2.0 Array 1434124_x_at - -
          Mouse Genome 430 2.0 Array 1417463_a_at - -
          Mouse Genome 430 2.0 Array 1434124_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66508_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66508_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700012429.mm.1 TC0700001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012429.mm.1 NM_025605.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012429.mm.1 NM_025605.3 Refseq
          Mouse Expression Set 430 1417463_PM_a_at - -
          Mouse Expression Set 430 1417463_a_at - -
          Mouse Expression Set 430 1427572_at - -
          Mouse Expression Set 430 1448718_at - -
          Mouse Expression Set 430 1434124_x_at - -
          Mouse Expression Set 430 1427572_PM_at - -
          Mouse Expression Set 430 1448718_PM_at - -
          Mouse Expression Set 430 1434124_PM_x_at - -
          Murine Genome U74v2 Set 96352_PM_at - -
          Murine Genome U74v2 Set 96352_at - -
          Murine Genome U74v2 Set 135142_PM_at - -
          Murine Genome U74v2 Set 135142_at - -
          Mouse Genome 430A 2.0 Array 1427572_at - -
          Mouse Genome 430A 2.0 Array 1417463_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427572_PM_at - -
          Mouse Genome 430A 2.0 Array 1417463_a_at - -
          Mouse Genome 430A 2.0 Array 1448718_PM_at - -
          Mouse Genome 430A 2.0 Array 1448718_at - -
          Mouse Genome 430A 2.0 Array 1434124_x_at - -
          Mouse Genome 430A 2.0 Array 1434124_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448718_at - -
          Affymetrix HT Mouse Genome 430A Array 1434124_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434124_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448718_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 351242 - -
          ABI Mouse Genome Survey Microarray 896928 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673380 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673380 - -
          mouse-6 v1.1 (Illumina) scl066508.5_236-S - -
          Illumina Sentrix mouse-6.v1 bead array 540279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066508.5_236-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2673380 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2673380 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066508.5_236-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline