Search
Search

Fcho2| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 24 - 25 | 113 | ||
| GenBank mRNA | AK031041.1 | - | 24 - 25 | 113 | ||
| AK142282.1 | - | 24 - 25 | 113 | |||
| AK161093.1 | - | 5 - 6 | 113 | |||
| AK164135.1 | - | 25 - 26 | 113 | |||
| AK169439.1 | - | 8 - 9 | 2005 | 113 | ||
| BC052456.1 | - | 24 - 25 | 6830607 | 113 | ||
Set Membership: |
|