Search
Search

Atp2b4| Gene Symbol: Atp2b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167949.2 | NP_001161421.1 | 12 - 13 | 2682 | 79 | |
| NM_213616.4 | NP_998781.1 | 12 - 13 | 2682 | 79 | ||
| GenBank mRNA | AY560895.1 | - | 12 - 13 | 2216 | 79 | |
| BC109172.1 | - | 12 - 13 | 2216 | 40048499 | 79 | |
| BC109173.1 | - | 12 - 13 | 2216 | 40048500 | 79 | |
| EU493094.2 | - | 12 - 13 | 2386 | 79 | ||
| EU493095.2 | - | 12 - 13 | 2386 | 79 | ||
| JF683384.1 | - | 12 - 13 | 2386 | 79 | ||
Set Membership: |
|