Search
Search
| Gene Symbol: Bok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016778.2 | NP_058058.1 | 4 - 5 | 87 | ||
| GenBank mRNA | AF027707.1 | - | 4 - 5 | 87 | ||
| AK156243.1 | - | 4 - 5 | 742 | 87 | ||
| AK168373.1 | - | 4 - 5 | 87 | |||
| AK171258.1 | - | 4 - 5 | 664 | 87 | ||
| BC030069.1 | - | 4 - 5 | 645 | 2936938 | 87 | |
Set Membership: |
|