Search
Search
| Gene Symbol: Pigr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011082.3 | NP_035212.2 | 6 - 7 | 1893 | 69 | |
| GenBank mRNA | AK128944.1 | - | 6 - 7 | 1893 | 69 | |
| AK142656.1 | - | 6 - 7 | 1895 | 69 | ||
| BC013556.1 | - | 6 - 7 | 1826 | 3601095 | 69 | |
| U06431.1 | - | 6 - 7 | 1797 | 69 | ||
Set Membership: |
|