Search
Search

Slco6c1| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028942.4 | NP_083218.1 | 10 - 11 | 1904 | 98 | |
| GenBank mRNA | AK016647.1 | - | 10 - 11 | 1852 | 98 | |
| AK029505.1 | - | 10 - 11 | 1905 | 98 | ||
| AK132668.1 | - | 10 - 11 | 1855 | 98 | ||
| AK161249.1 | - | 10 - 11 | 1855 | 98 | ||
| BC139066.1 | - | 10 - 11 | 1807 | 8862088 | 98 | |
| BC139067.1 | - | 10 - 11 | 1807 | 8862089 | 98 | |
Set Membership: |
|