Search
Search

Gabra3| Gene Symbol: Gabra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008067.4 | NP_032093.3 | 6 - 7 | 903 | 125 | |
| GenBank mRNA | AK039144.1 | - | 6 - 7 | 844 | 125 | |
| AK134760.1 | - | 6 - 7 | 812 | 125 | ||
| BC119317.1 | - | 6 - 7 | 701 | 8734066 | 125 | |
| BC120686.1 | - | 6 - 7 | 701 | 40129609 | 125 | |
| M86568.1 | - | 6 - 7 | 805 | 125 | ||
Set Membership: |
|