Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01294506_g1
          See other 4933406C10RIK GE Assays ›
          Gene Symbol:
          4933406C10Rik
          Gene Name:
          RIKEN cDNA 4933406C10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 32919384 - 32953789 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01294506_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933406C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_044986.1 5 - 6 837 76
          GenBank mRNA AK041289.1 - 5 - 6 838 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nicotinamide phosphoribosyltransferase Nampt 1110035O14Rik,
          AI314458,
          AI480535,
          NAmPRTase,
          Pbef,
          Pbef1,
          Visfatin
          -66.0kb Anti-sense Chr. 12 - 32820335 - 32853369 on NCBI Build GRCm38
          Downstream synaptophysin-like protein Sypl AI314763,
          AI604763,
          D12Ertd446e,
          PanI,
          Pphn,
          Sypl1
          33.1kb Anti-sense Chr. 12 - 32953873 - 32979502 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001613.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2168118 AK041289.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868589 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32897 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168118 AK041289.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07581194 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02487055 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02487020 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07581159 - -
          Whole Mouse Genome G4122A A_52_P425317 AK016682.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02487055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09826199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10100293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09955220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09798582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10174460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09955225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09798586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10164195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10347214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28338902 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430246_PM_at - -
          Mouse Genome 430 2.0 Array 1430246_at - -
          Mouse Genome 430 2.0 Array 1430246_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200011220.mm.1 NR_044986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011220.mm.1 NR_044986.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011220.mm.1 TC1200001613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011219.mm.1 NR_044986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011219.mm.1 NR_044986.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011219.mm.1 TC1200001613.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200011221.mm.1 NR_044986.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011221.mm.1 NR_044986.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200011221.mm.1 TC1200001613.mm.1 MTA TC
          Mouse Expression Set 430 1430246_at - -
          Mouse Expression Set 430 1430246_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430246_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430246_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 309783 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline