Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01295205_m1
          See other ZFP879 GE Assays ›
          Gene Symbol:
          Zfp879
          Gene Name:
          zinc finger protein 879
          Gene Aliases:
          Znf879
          Chromosome Location:
          Chr.11: 50832031 - 50841563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01295205_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp879
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290779.1 NP_001277708.1 3 - 4 342 68
          NM_173387.3 NP_775563.1 4 - 5 469 68
          GenBank mRNA AK036152.1 - 4 - 5 470 68
          BC118537.1 - 3 - 4 336 40094138 68

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC118537.1, NM_001290779.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp879

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790926 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035549 NM_173387.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035549 AK036152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169019 BC118537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169019 NM_173387.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169019 AK036152.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65392 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166865 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169019 BC118537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169019 NM_173387.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169019 AK036152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035549 NM_173387.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035549 AK036152.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27790885 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02323017 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166865 - -
          Whole Mouse Genome G4122A A_51_P508683 BC118537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508683 NM_173387.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P508683 AK036152.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124960 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108813 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09745098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09745102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09745087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09745103 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790927 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27790925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09081002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09745107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09338131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09745089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09745106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09019543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09745084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09077224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09015220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09019542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09710689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09078905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790852 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385645 - -
          Affymetrix HT MG-430 PM Array Plate 1441207_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385642 - -
          Mouse Genome 430 2.0 Array 1441207_PM_at - -
          Mouse Genome 430 2.0 Array 1441207_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214779_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214779_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100024456.mm.1 NM_173387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024456.mm.1 TC1100002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024455.mm.1 NM_173387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024455.mm.1 TC1100002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024454.mm.1 NM_173387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024454.mm.1 TC1100002680.mm.1 MTA TC
          Mouse Expression Set 430 1441207_at - -
          Mouse Expression Set 430 1441207_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441207_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441207_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 784635 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2680020 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680020 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2749383 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2749383 - -
          mouse-6 v1.1 (Illumina) scl40268.5.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680020 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40268.5.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680020 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2749383 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2749383 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline