Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01299396_m1
          See other DNAJC21 GE Assays ›
          Gene Symbol:
          Dnajc21
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 21
          Gene Aliases:
          4930461P20Rik, 9930116P15Rik
          Chromosome Location:
          Chr.15: 10446763 - 10470516 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01299396_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030046.2 NP_084322.2 5 - 6 898 61
          GenBank mRNA AK141303.1 - 2 - 3 358 61
          BC048354.1 - 1 - 2 189 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001201.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150389 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150389 AK140224.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007165 AK141303.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 NM_030046.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853191 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139519 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150389 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150389 AK140224.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007165 AK141303.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 AK020688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 NM_030046.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02972427 - -
          Whole Mouse Genome G4122A A_51_P439857 NM_030046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 AK020688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 NM_030046.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P439857 AK020688.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12433953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12278104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12155530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11840635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11980289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11918821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12093895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12034088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11878832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11886250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30010662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11981071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10427709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10427715 - -
          Affymetrix HT MG-430 PM Array Plate 1460711_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420221_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420220_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1457281_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420219_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10427704 - -
          Mouse Genome 430 2.0 Array 1460711_PM_at - -
          Mouse Genome 430 2.0 Array 1420219_at - -
          Mouse Genome 430 2.0 Array 1420219_PM_at - -
          Mouse Genome 430 2.0 Array 1420220_PM_x_at - -
          Mouse Genome 430 2.0 Array 1460711_at - -
          Mouse Genome 430 2.0 Array 1420221_PM_at - -
          Mouse Genome 430 2.0 Array 1420220_x_at - -
          Mouse Genome 430 2.0 Array 1420221_at - -
          Mouse Genome 430 2.0 Array 1457281_PM_at - -
          Mouse Genome 430 2.0 Array 1457281_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78244_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78244_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500009667.mm.1 NM_030046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009667.mm.1 NM_030046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500009667.mm.1 TC1500001201.mm.1 MTA TC
          Mouse Expression Set 430 1460711_at - -
          Mouse Expression Set 430 1460711_PM_at - -
          Mouse Expression Set 430 1420220_PM_x_at - -
          Mouse Expression Set 430 1420220_x_at - -
          Mouse Expression Set 430 1420221_at - -
          Mouse Expression Set 430 1420219_at - -
          Mouse Expression Set 430 1420219_PM_at - -
          Mouse Expression Set 430 1420221_PM_at - -
          Mouse Expression Set 430 1457281_at - -
          Mouse Expression Set 430 1457281_PM_at - -
          Murine Genome U74v2 Set 93478_at - -
          Murine Genome U74v2 Set 93478_PM_at - -
          Murine Genome U74v2 Set 169515_r_at - -
          Murine Genome U74v2 Set 134385_at - -
          Murine Genome U74v2 Set 134385_PM_at - -
          Murine Genome U74v2 Set 169515_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1460711_at - -
          Mouse Genome 430A 2.0 Array 1460711_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460711_at - -
          Affymetrix HT Mouse Genome 430A Array 1460711_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1420221_at - -
          Affymetrix HT Mouse Genome 430B Array 1457281_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1420219_at - -
          Affymetrix HT Mouse Genome 430B Array 1420220_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1420220_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1420219_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1420221_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457281_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 678695 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline