Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01301785_m1
          See other CCR7 GE Assays ›
          Gene Symbol:
          Ccr7
          Gene Name:
          chemokine (C-C motif) receptor 7
          Gene Aliases:
          CC-CKR-7, CCR-7, CD197, Cdw197, Cmkbr7, EBI1, Ebi1h
          Chromosome Location:
          Chr.11: 99144199 - 99155077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01301785_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccr7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007719.2 NP_031745.2 2 - 3 140 78
          GenBank mRNA AK037965.1 - 2 - 3 141 78
          AK154349.1 - 2 - 3 141 78
          AK154736.1 - 2 - 3 141 78
          AK154849.1 - 2 - 3 141 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003696.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus Mouse Chemokine Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccr7

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of T cell polarity
          myeloid dendritic cell chemotaxis
          lymphocyte chemotaxis across high endothelial venule
          positive regulation of dendritic cell antigen processing and presentation
          regulation of tolerance induction to self antigen
          positive regulation of hypersensitivity
          positive regulation of humoral immune response
          chemotaxis
          inflammatory response
          immune response
          signal transduction
          G-protein coupled receptor signaling pathway
          activation of JUN kinase activity
          positive regulation of macrophage chemotaxis
          positive regulation of T cell chemotaxis
          actin cytoskeleton organization
          response to lipopolysaccharide
          regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          positive regulation of tumor necrosis factor production
          response to prostaglandin E
          chemokine (C-C motif) ligand 19 signaling pathway
          chemokine (C-C motif) ligand 21 signaling pathway
          positive regulation of T cell proliferation
          positive regulation of GTPase activity
          negative thymic T cell selection
          positive regulation of T-helper 1 cell differentiation
          positive regulation of receptor-mediated endocytosis
          lymph node development
          regulation of interleukin-1 beta secretion
          positive regulation of interleukin-1 beta secretion
          positive regulation of phagocytosis
          positive regulation of T cell receptor signaling pathway
          positive regulation of filopodium assembly
          chemokine-mediated signaling pathway
          cellular response to cytokine stimulus
          response to nitric oxide
          interleukin-12 secretion
          positive regulation of neutrophil chemotaxis
          lymphocyte migration into lymph node
          mature conventional dendritic cell differentiation
          negative regulation of leukocyte apoptotic process
          positive regulation of cell motility
          positive regulation of thymocyte migration
          positive regulation of dendritic cell chemotaxis
          positive regulation of immunological synapse formation
          positive regulation of T cell costimulation
          positive regulation of glycoprotein biosynthetic process involved in immunological synapse formation
          regulation of dendritic cell dendrite assembly
          signal transducer activity
          G-protein coupled receptor activity
          chemokine receptor activity
          protein binding
          C-C chemokine receptor activity
          chemokine (C-C motif) ligand 19 binding
          chemokine (C-C motif) ligand 21 binding
          C-C motif chemokine 19 receptor activity
          C-C motif chemokine 21 receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100034090.mm.1 NM_007719.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034090.mm.1 NM_007719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100034090.mm.1 TC1100003696.mm.1 MTA TC

          Back To Top
           

          Citations (1)

          Requirement for T-bet in the aberrant differentiation of unhelped memory CD8+ T cells.
          Authors
          Intlekofer AM, Takemoto N, Kao C, Banerjee A, Schambach F, Northrop JK, Shen H, Wherry EJ, Reiner SL
          Journal
          J Exp Med - Page No 2015 (Mon Sep 03 00:00:00 EDT 2007)
          Volume (Year)
          204 (2007)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline