Search
Search
| Gene Symbol: Cdc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025779.1 | NP_001020950.1 | 2 - 3 | 291 | 74 | |
| NM_011799.2 | NP_035929.1 | 2 - 3 | 367 | 74 | ||
| GenBank mRNA | AJ009559.1 | - | 2 - 3 | 384 | 74 | |
| AJ223087.1 | - | 2 - 3 | 369 | 74 | ||
| AK012192.1 | - | 2 - 3 | 321 | 74 | ||
| AK075975.1 | - | 2 - 3 | 321 | 73 | ||
| AK085020.1 | - | 2 - 3 | 321 | 74 | ||
| AK162669.1 | - | 2 - 3 | 332 | 74 | ||
| AK166003.1 | - | 2 - 3 | 372 | 74 | ||
| BC052434.1 | - | 2 - 3 | 291 | 6837113 | 74 | |
Set Membership: |
|