Search
Search

Clcn7| Gene Symbol: Clcn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011930.3 | NP_036060.1 | 19 - 20 | 1871 | 59 | |
| GenBank mRNA | AK030444.1 | - | 19 - 20 | 1872 | 59 | |
| AK032674.1 | - | 6 - 7 | 601 | 59 | ||
| AK150395.1 | - | 19 - 20 | 1782 | 59 | ||
| AK190848.1 | - | 1 - 2 | 67 | 59 | ||
| BC050907.1 | - | 19 - 20 | 1843 | 5698121 | 59 | |
| BC053049.1 | - | 19 - 20 | 1843 | 5698121 | 59 | |
| BC054799.1 | - | 19 - 20 | 1851 | 6849205 | 59 | |
Set Membership: |
|