Search
Search

Ttll1| Gene Symbol: Ttll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178869.4 | NP_849200.2 | 8 - 9 | 1132 | 71 | |
| GenBank mRNA | AK031917.1 | - | 8 - 9 | 1103 | 71 | |
| AK047046.1 | - | 8 - 9 | 1117 | 71 | ||
| AK084005.1 | - | 8 - 9 | 1100 | 71 | ||
| AK140472.1 | - | 8 - 9 | 1100 | 71 | ||
| AK168786.1 | - | 8 - 9 | 1103 | 71 | ||
| AK169150.1 | - | 8 - 9 | 1119 | 71 | ||
| BC010510.1 | - | 8 - 9 | 1081 | 4191464 | 71 | |
Set Membership: |
|