Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01313840_g1
          See other MYH6 GE Assays ›
          Gene Symbol:
          Myh6
          Gene Name:
          myosin, heavy polypeptide 6, cardiac muscle, alpha
          Gene Aliases:
          A830009F23Rik, AA517445, Myhc-a, Myhca, alpha-MHC
          Chromosome Location:
          Chr.14: 54941921 - 54968014 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01313840_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164171.1 NP_001157643.1 33 - 34 5156 80
          NM_010856.4 NP_034986.1 33 - 34 5092 80
          GenBank mRNA BC110700.1 - 33 - 34 5138 6786621 80
          BC138027.1 - 32 - 33 5031 8861046 80
          M74751.1 - 82
          M76599.1 - 31 - 32 4954 80
          M76600.1 - 31 - 32 4954 80
          M76601.1 - 33 - 34 5056 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002073.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myh6

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of the force of heart contraction
          regulation of heart rate
          muscle contraction
          striated muscle contraction
          adult heart development
          visceral muscle development
          regulation of heart contraction
          metabolic process
          regulation of blood pressure
          actin filament-based movement
          muscle filament sliding
          myofibril assembly
          BMP signaling pathway
          regulation of ATPase activity
          sarcomere organization
          ATP metabolic process
          cardiac muscle fiber development
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          canonical Wnt signaling pathway
          regulation of heart growth
          microfilament motor activity
          nucleotide binding
          motor activity
          actin binding
          protein binding
          calmodulin binding
          ATP binding
          ATPase activity
          protein kinase binding
          actin-dependent ATPase activity
          calcium-dependent ATPase activity
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400015679.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015679.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015679.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015679.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015679.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015677.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015677.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015677.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015677.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015677.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015681.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015681.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015681.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015681.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015681.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015682.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015682.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015682.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015682.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015682.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015678.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015678.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015678.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015678.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015678.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015680.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015680.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015680.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015680.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015680.mm.1 TC1400002073.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline