Search
Search
| Gene Symbol: Fhit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF047699.1 | - | 3 - 4 | 450 | 74 | |
| AF055573.1 | - | 316 | 74 | |||
| BC012662.1 | - | 1 - 2 | 175 | 4036815 | 74 | |
| CJ053929.1 | - | 209 | 74 | |||
| CJ056464.1 | - | 4 - 5 | 338 | 74 | ||
| CJ058331.1 | - | 4 - 5 | 336 | 74 | ||
Set Membership: |
|