Search
Search
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276387.1 | NP_001263316.1 | 10 - 11 | 1855 | 66 | |
| NM_010095.6 | NP_034225.1 | 10 - 11 | 1850 | 66 | ||
| GenBank mRNA | AK053013.1 | - | 10 - 11 | 1851 | 66 | |
| AK139262.1 | - | 10 - 11 | 1562 | 66 | ||
| BC049188.1 | - | 9 - 10 | 1149 | 6312260 | 66 | |
| BC050922.1 | - | 9 - 10 | 1372 | 6409737 | 66 | |
| U71189.1 | - | 9 - 10 | 1035 | 66 | ||
| U82441.1 | - | 9 - 10 | 904 | 66 | ||
| U92703.1 | - | 9 - 10 | 1255 | 66 | ||
Set Membership: |
|