Search
Search

Ccdc61| Gene Symbol: Ccdc61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033314.3 | NP_001028486.1 | 12 - 13 | 1643 | 81 | |
| GenBank mRNA | AK082980.1 | - | 12 - 13 | 1597 | 81 | |
Set Membership: |
|