Search
Search

Gpnmb| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053110.4 | NP_444340.3 | 6 - 7 | 1275 | 67 | |
| GenBank mRNA | AF322054.1 | - | 6 - 7 | 1113 | 67 | |
| AJ251685.1 | - | 6 - 7 | 1160 | 67 | ||
| AK044764.1 | - | 6 - 7 | 1169 | 67 | ||
| AK076347.1 | - | 6 - 7 | 1144 | 67 | ||
| AK079220.1 | - | 1117 | 67 | |||
| AK149740.1 | - | 6 - 7 | 1183 | 67 | ||
| AK150739.1 | - | 6 - 7 | 1144 | 67 | ||
| AK154617.1 | - | 6 - 7 | 1179 | 67 | ||
| AK159165.1 | - | 6 - 7 | 1174 | 67 | ||
| AK159270.1 | - | 6 - 7 | 1182 | 67 | ||
| AK159747.1 | - | 6 - 7 | 1144 | 67 | ||
| AK159908.1 | - | 6 - 7 | 1150 | 67 | ||
| AK170506.1 | - | 6 - 7 | 1174 | 67 | ||
| AK171441.1 | - | 6 - 7 | 1144 | 67 | ||
| AK171465.1 | - | 6 - 7 | 1144 | 67 | ||
| AK171474.1 | - | 6 - 7 | 1144 | 67 | ||
| AK171517.1 | - | 6 - 7 | 1262 | 67 | ||
| AK171608.1 | - | 6 - 7 | 1144 | 67 | ||
| AK171618.1 | - | 6 - 7 | 1144 | 67 | ||
| BC026375.1 | - | 6 - 7 | 1149 | 4164706 | 67 | |
Set Membership: |
|