Search
Search
| Gene Symbol: Enam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017468.3 | NP_059496.1 | 2 - 3 | 274 | 93 | |
| GenBank mRNA | BC115848.1 | - | 2 - 3 | 186 | 40048943 | 93 |
| U82698.1 | - | 2 - 3 | 275 | 93 | ||
Set Membership: |
|