Search
Search

Hdac1| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008228.2 | NP_032254.1 | 12 - 13 | 1391 | 125 | |
| GenBank mRNA | AK076080.1 | - | 12 - 13 | 1373 | 125 | |
| AK194630.1 | - | 2 - 3 | 125 | |||
| BC092070.1 | - | 12 - 13 | 1386 | 4217199 | 125 | |
| BC108371.1 | - | 12 - 13 | 1392 | 4976514 | 125 | |
| U80780.1 | - | 12 - 13 | 1388 | 125 | ||
| X98207.1 | - | 12 - 13 | 1391 | 125 | ||
Set Membership: |
|