Search
Search

C1qtnf9| Gene Symbol: C1qtnf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183175.4 | NP_898998.2 | 3 - 4 | 287 | 97 | |
| GenBank mRNA | BC115632.1 | - | 3 - 4 | 272 | 40102920 | 97 |
| BC115633.1 | - | 3 - 4 | 272 | 40102924 | 97 | |
| DQ002401.1 | - | 3 - 4 | 287 | 97 | ||
Set Membership: |
|