Search
Search

Inhba| Gene Symbol: Inhba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008380.1 | NP_032406.1 | 1 - 2 | 61 | 154 | |
| GenBank mRNA | AK134991.1 | - | 3 - 4 | 512 | 154 | |
| X69619.1 | - | 1 - 2 | 61 | 154 | ||
Set Membership: |
|