Search
Search

Pcdhb1| Gene Symbol: Pcdhb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053126.1 | NP_444356.1 | 1 | 2331 | 111 | |
| GenBank mRNA | AY013770.1 | - | 1 | 2331 | 111 | |
| BC132112.1 | - | 1 | 2391 | 40130389 | 111 | |
| BC132296.1 | - | 1 | 2391 | 40130573 | 111 | |
Set Membership: |
|