Search
Search
| Gene Symbol: Oog3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201258.2 | NP_957710.2 | 1 - 2 | 65 | 112 | |
| GenBank mRNA | AK136030.1 | - | 1 - 2 | 66 | 112 | |
| AY573563.1 | - | 1 - 2 | 62 | 112 | ||
| BC053700.1 | - | 1 - 2 | 62 | 30049894 | 112 | |
Set Membership: |
|