Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03806928_gH
          See other MUP1 GE Assays ›
          Gene Symbol:
          Mup1 Gm2083 LOC100048884 Mup10 Mup11 Mup12 Mup14 Mup15 Mup16 Mup17 Mup19 Mup2 Mup7 Mup8 Mup9
          Gene Name:
          major urinary protein 1
          major urinary protein LOC100048885
          novel member of the major urinary protein (Mup) gene family
          major urinary protein 10
          major urinary protein 11
          major urinary protein 12
          major urinary protein 14
          major urinary protein 15
          major urinary protein 16
          major urinary protein 17
          major urinary protein 19
          major urinary protein 2
          major urinary protein 7
          major urinary protein 8
          major urinary protein 9
          Gene Aliases:
          Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
          -
          Mup6
          2610016E04Rik, Mup8
          Gm12549, OTTMUSG00000007431
          Gm2024
          Gm13514, Mup12, OTTMUSG00000012493
          Gm2068, Mup13
          -
          Gm12557, MUP 17, OTTMUSG00000007480
          100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
          AA589603, Mup-2, Mup4
          Gm12546, Mup3, OTTMUSG00000007428
          Gm12809, Mup5, OTTMUSG00000008509
          Gm14076, Mup2, Mup6, OTTMUSG00000015595
          Chromosome Location:
          Chr.4: 60498012 - 60501960 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.4: 61670177 - 61674098 on Build GRCm38
          Chromosome Location:
          Chr.4: 60578260 - 60582155 on Build GRCm38
          Chromosome Location:
          Chr.4: 60658466 - 60662395 on Build GRCm38
          Chromosome Location:
          Chr.4: 60737381 - 60741281 on Build GRCm38
          Chromosome Location:
          Chr.4: 61300037 - 61303998 on Build GRCm38
          Chromosome Location:
          Chr.4: 61435787 - 61439714 on Build GRCm38
          Chromosome Location:
          Chr.4: 61515592 - 61519503 on Build GRCm38
          Chromosome Location:
          Chr.4: 61591929 - 61595853 on Build GRCm38
          Chromosome Location:
          Chr.4: 61778324 - 61782236 on Build GRCm38
          Chromosome Location:
          Chr.4: 60135913 - 60139872 on Build GRCm38
          Chromosome Location:
          Chr.4: 60066469 - 60070481 on Build GRCm38
          Chromosome Location:
          Chr.4: 60218620 - 60222599 on Build GRCm38
          Chromosome Location:
          Chr.4: 60418046 - 60421961 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          Assay ID Mm03806928_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mup1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163010.1 NP_001156482.1 3 - 4 412 128
          NM_001163011.1 NP_001156483.1 3 - 4 412 128
          NM_031188.2 NP_112465.2 3 - 4 412 128
          GenBank mRNA BC012221.1 - 3 - 4 358 4195753 128
          BC013649.1 - 346 4195401 128
          BC037152.1 - 236 4161709 128
          BC091744.1 - 349 5102271 128
          BC091775.1 - 3 - 4 359 5137341 128
          BC092254.1 - 346 5100458 128
          BC093515.1 - 346 128
          BC099597.1 - 349 947916 128
          BC100586.1 - 366 6769126 128
          BC106092.1 - 3 - 4 349 5098340 128
          M16355.1 - 3 - 4 412 128
          M27609.1 - 214 128
          X03524.1 - 214 128
          X04115.1 - 170 128
          Gene Symbol: Gm2083
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134644.1 NP_001128116.1 3 - 4 366 128
          Gene Symbol: LOC100048884
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199333.1 NP_001186262.1 3 - 4 373 128
          GenBank mRNA BC055353.1 - 348 128
          Gene Symbol: Mup10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122647.1 NP_001116119.1 3 - 4 368 128
          GenBank mRNA AJ309921.1 - 301 128
          AK011413.1 - 1 - 2 420 128
          CB948918.1 - 277 128
          EU882231.1 - 3 - 4 301 128
          Gene Symbol: Mup11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164526.1 NP_001157998.1 399 128
          GenBank mRNA AK013972.1 - 3 - 4 376 128
          Gene Symbol: Mup12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199995.1 NP_001186924.1 3 - 4 367 128
          Gene Symbol: Mup14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199999.1 NP_001186928.1 3 - 4 410 128
          Gene Symbol: Mup15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001200004.1 NP_001186933.1 3 - 4 373 128
          Gene Symbol: Mup16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199936.1 NP_001186865.1 3 - 4 367 128
          GenBank mRNA AW112504.1 - 4 - 5 128
          Gene Symbol: Mup17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001200006.1 NP_001186935.1 3 - 4 373 128
          Gene Symbol: Mup19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135127.2 NP_001128599.1 3 - 4 367 128
          GenBank mRNA AK013259.1 - 3 - 4 367 128
          EU882232.1 - 3 - 4 295 128
          Gene Symbol: Mup2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045550.2 NP_001039015.1 3 - 4 371 128
          NM_001286096.1 NP_001273025.1 2 - 3 237 128
          NM_008647.4 NP_032673.3 3 - 4 371 128
          GenBank mRNA AW112687.1 - 3 - 4 128
          BC012259.1 - 360 4196403 128
          BC059097.1 - 346 4159984 128
          CB593503.1 - 369 128
          M16356.1 - 359 128
          Gene Symbol: Mup7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134675.1 NP_001128147.1 3 - 4 476 128
          GenBank mRNA EU882230.1 - 3 - 4 301 128
          Gene Symbol: Mup8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134676.1 NP_001128148.1 3 - 4 431 128
          Gene Symbol: Mup9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281979.1 NP_001268908.1 3 - 4 367 128
          GenBank mRNA BI219655.1 - 3 - 4 348 128
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BC094452.1 - 347 Chr. - - [-] on Build 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          transport
          pheromone activity
          pheromone binding
          molecular_function
          transporter activity
          small molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400022538.mm.1 TC0400004211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022538.mm.1 NM_001045550.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022538.mm.1 NM_001164526.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022538.mm.1 NM_001164526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022538.mm.1 NM_008647.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022621.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022621.mm.1 NM_001199999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022621.mm.1 NM_001199999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022636.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022636.mm.1 NM_001200004.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022636.mm.1 NM_001286096.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022624.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022624.mm.1 NM_001199999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022624.mm.1 NM_001199999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022485.mm.1 TC0400004210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022485.mm.1 NM_001134676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022485.mm.1 NM_001134676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022620.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022620.mm.1 NM_001199999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022620.mm.1 NM_001199999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022620.mm.1 NM_001286096.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022634.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022634.mm.1 NM_001200004.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022634.mm.1 NM_001286096.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022571.mm.1 TC0400004213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022487.mm.1 TC0400004210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022487.mm.1 NM_001134676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022487.mm.1 NM_001134676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022488.mm.1 TC0400004210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022488.mm.1 NM_001134676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022488.mm.1 NM_001134676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022537.mm.1 TC0400004211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022537.mm.1 NM_001045550.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022537.mm.1 NM_001164526.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022537.mm.1 NM_001164526.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022537.mm.1 NM_008647.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022626.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022626.mm.1 NM_001199999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022626.mm.1 NM_001199999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022658.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022658.mm.1 NM_001200006.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022658.mm.1 NM_001200006.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022688.mm.1 TC0400002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022688.mm.1 NM_001135127.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022688.mm.1 NM_001135127.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022452.mm.1 TC0400004208.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022452.mm.1 NM_001134675.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022452.mm.1 NM_001134675.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022500.mm.1 TC0400004210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022500.mm.1 NM_001281979.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022522.mm.1 TC0400004211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022522.mm.1 NM_001122647.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022522.mm.1 NM_001122647.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022469.mm.1 TC0400004209.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022623.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022623.mm.1 NM_001199999.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022623.mm.1 NM_001199999.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022645.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022645.mm.1 NM_001199936.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022645.mm.1 NM_001199936.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022553.mm.1 TC0400004212.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022553.mm.1 NM_001199995.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022553.mm.1 NM_001199995.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022672.mm.1 TC0400002769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022672.mm.1 NM_001199333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022672.mm.1 NM_001199333.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline