Search
Search
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052041.1 | - | 1 | 1450 | 74 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DIP2 disco-interacting protein 2 homolog A (Drosophila) | Dip2a |
4931420H10Rik, AI426328, Dip2, Kiaa0184-hp, mKIAA0184 |
-6.0kb | Sense | Chr. 10 - 76263049 - 76345301 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700094J05 gene | 1700094J05Rik | 35.5kb | Anti-sense | Chr. 10 - 76389015 - 76401507 on NCBI Build GRCm38 |
Set Membership: |
|