Search
Search
| Gene Symbol: Ybey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004411.1 | - | 1 | 250 | 88 | |
| AK051909.1 | - | 1 | 286 | 88 | ||
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2610028H24 gene | 2610028H24Rik | ORF67 | -7.3kb | Anti-sense | Chr. 10 - 76449023 - 76461218 on NCBI Build GRCm38 |
| Downstream | minichromosome maintenance deficient 3 (S. cerevisiae) associated protein | Mcm3ap |
GANP, mKIAA0572 |
144.0 | Anti-sense | Chr. 10 - 76468971 - 76515859 on NCBI Build GRCm38 |
Set Membership: |
|