Search
Search

Trim46| Gene Symbol: Trim46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121683.1 | 7 - 8 | 1532 | 112 | ||
| GenBank mRNA | BC065049.1 | - | 7 - 8 | 1445 | 6418851 | 112 |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
mucin 1, transmembrane |
Muc1 |
CD227, EMA, Muc-1 |
-5.3kb | Anti-sense | Chr. 3 - 89229056 - 89233381 on NCBI Build GRCm38 |
| Downstream | keratinocyte associated protein 2 | Krtcap2 |
0610010I12Rik, Kcp2 |
7.3kb | Anti-sense | Chr. 3 - 89246300 - 89249723 on NCBI Build GRCm38 |
Set Membership: |
|