Search
Search
| Gene Symbol: RHOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127285.1 | NP_001120757.1 | 1 - 2 | 148 | 65 | |
| XM_012097249.3 | XP_011952639.1 | 4 - 5 | 7135 | 65 | ||
| XM_012097252.3 | XP_011952642.1 | 3 - 4 | 7099 | 65 | ||
| XM_012097256.3 | XP_011952646.1 | 4 - 5 | 6887 | 65 | ||
| XM_012097257.3 | XP_011952647.1 | 4 - 5 | 7247 | 65 | ||
| XM_012097260.3 | XP_011952650.1 | 6 - 7 | 3712 | 65 | ||
| XM_027980221.1 | XP_027836022.1 | 4 - 5 | 5430 | 65 | ||
| XM_027980222.1 | XP_027836023.1 | 4 - 5 | 4273 | 65 | ||
| XM_027980223.1 | XP_027836024.1 | 5 - 6 | 3796 | 65 | ||
| GenBank mRNA | EU626204.1 | - | 1 - 2 | 148 | 65 | |
Set Membership: |
|