Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Oa04733236_m1
          See other TCF4 GE Assays ›
          Gene Symbol:
          TCF4
          Gene Name:
          transcription factor 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.23: 60572934 - 60957822 on Build Oar_rambouillet_v1.0
          Species:
          Sheep
          Species Specific ID (Flybase ID):
          -
          Assay ID Oa04733236_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TCF4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_004020558.4 XP_004020607.1 4 - 5 3265 50
          XM_012103755.3 XP_011959145.1 12 - 13 1494 50
          XM_012103763.3 XP_011959153.1 11 - 12 986 50
          XM_012103766.3 XP_011959156.1 4 - 5 3285 50
          XM_012103767.3 XP_011959157.1 4 - 5 3286 50
          XM_012103768.3 XP_011959158.1 11 - 12 1458 50
          XM_015103770.2 XP_014959256.1 11 - 12 1013 50
          XM_015103771.2 XP_014959257.1 9 - 10 833 50
          XM_015103772.2 XP_014959258.1 9 - 10 833 50
          XM_015103774.2 XP_014959260.1 6 - 7 956 50
          XM_015103776.2 XP_014959262.1 6 - 7 783 50
          XM_015103777.2 XP_014959263.1 6 - 7 953 50
          XM_027960898.1 XP_027816699.1 10 - 11 878 50
          XM_027960899.1 XP_027816700.1 10 - 11 881 50
          XM_027960900.1 XP_027816701.1 10 - 11 881 50
          XM_027960901.1 XP_027816702.1 10 - 11 878 50
          XM_027960902.1 XP_027816703.1 9 - 10 845 50
          XM_027960904.1 XP_027816705.1 9 - 10 848 50
          XM_027960905.1 XP_027816706.1 8 - 9 701 50
          XM_027960906.1 XP_027816707.1 12 - 13 1046 50
          XM_027960907.1 XP_027816708.1 11 - 12 1013 50
          XM_027960908.1 XP_027816709.1 11 - 12 1010 50
          XM_027960909.1 XP_027816710.1 11 - 12 1013 50
          XM_027960910.1 XP_027816711.1 6 - 7 1093 50
          XM_027960911.1 XP_027816712.1 4 - 5 3291 50
          XM_027960912.1 XP_027816713.1 4 - 5 543 50
          XM_027960913.1 XP_027816714.1 4 - 5 568 50
          XM_027960914.1 XP_027816715.1 6 - 7 661 50

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Ovis aries

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline