Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02830876_m1
See other ABLIM1 GE Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 114949909 - 115288826 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02830876_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015148232.2
XP_015003718.2
2601
92
XM_015148233.2
XP_015003719.2
2595
92
XM_015148234.2
XP_015003720.1
2753
92
XM_015148235.2
XP_015003721.1
2033
92
XM_015148236.2
XP_015003722.2
2517
92
XM_015148237.2
XP_015003723.2
2486
92
XM_015148238.2
XP_015003724.2
2475
92
XM_015148239.2
XP_015003725.2
2454
92
XM_015148240.2
XP_015003726.2
2391
92
XM_015148241.2
XP_015003727.2
2376
92
XM_015148242.2
XP_015003728.2
2370
92
XM_015148243.2
XP_015003729.2
2334
92
XM_015148245.2
XP_015003731.1
2015
92
XM_015148247.2
XP_015003733.2
2292
92
XM_015148248.2
XP_015003734.2
2286
92
XM_015148249.2
XP_015003735.2
2250
92
XM_015148250.2
XP_015003736.2
2229
92
XM_015148251.2
XP_015003737.1
1981
92
XM_015148252.2
XP_015003738.2
2145
92
XM_015148253.2
XP_015003739.1
1283
92
XM_015148254.2
XP_015003740.1
2035
92
XM_028826890.1
XP_028682723.1
2397
92
XM_028826891.1
XP_028682724.1
2370
92
XM_028826892.1
XP_028682725.1
1556
92
XM_028826893.1
XP_028682726.1
2056
92
XM_028826894.1
XP_028682727.1
1978
92
XM_028826895.1
XP_028682728.1
1972
92
XM_028826896.1
XP_028682729.1
1936
92
XM_028826897.1
XP_028682730.1
1831
92
XM_028826898.1
XP_028682731.1
1930
92
GenBank mRNA
JV468378.1
-
330
92
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Recommended Assay
Macaca mulatta
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline