Search
Search

SNCG| Gene Symbol: SNCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266560.1 | NP_001253489.1 | 458 | 112 | ||
| XM_028853290.1 | XP_028709123.1 | 540 | 112 | |||
| XM_028853291.1 | XP_028709124.1 | 425 | 112 | |||
| XM_028853292.1 | XP_028709125.1 | 425 | 112 | |||
| XM_028853293.1 | XP_028709126.1 | 425 | 112 | |||
| GenBank mRNA | JU323203.1 | - | 458 | 112 | ||
| JV479476.1 | - | 252 | 112 | |||
Set Membership: |
|