Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07221133_g1
See other ADGRL2 GE Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
LPHN2
Chromosome Location:
Chr.1: 142429463 - 143111519 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07221133_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015143262.2
XP_014998748.2
2324
91
XM_015143264.2
XP_014998750.2
1704
91
XM_015143272.2
XP_014998758.2
1736
91
XM_015143279.2
XP_014998765.2
2321
91
XM_015143285.2
XP_014998771.2
2309
91
XM_015143290.2
XP_014998776.2
1689
91
XM_015143297.2
XP_014998783.2
2324
91
XM_015143308.2
XP_014998794.2
2322
91
XM_015143318.2
XP_014998804.2
2324
91
XM_015143325.2
XP_014998811.2
2323
91
XM_015143330.2
XP_014998816.2
2324
91
XM_015143342.2
XP_014998828.2
2321
91
XM_015143348.2
XP_014998834.2
2323
91
XM_015143354.2
XP_014998840.2
2320
91
XM_015143359.2
XP_014998845.2
2308
91
XM_015143365.2
XP_014998851.2
2307
91
XM_015143369.2
XP_014998855.2
2320
91
XM_015143370.2
XP_014998856.2
2323
91
XM_015143371.2
XP_014998857.2
2321
91
XM_015143385.2
XP_014998871.2
2320
91
XM_015143389.2
XP_014998875.2
2322
91
XM_015143397.2
XP_014998883.2
2319
91
XM_015143404.2
XP_014998890.2
2322
91
XM_015143411.2
XP_014998897.2
2319
91
XM_015143418.2
XP_014998904.2
2306
91
XM_015143426.2
XP_014998912.2
1702
91
XM_015143428.2
XP_014998914.2
2330
91
XM_015143437.2
XP_014998923.2
2328
91
XM_015143444.2
XP_014998930.2
2328
91
XM_015143451.2
XP_014998937.2
2328
91
XM_015143466.2
XP_014998952.2
2327
91
XM_015143470.2
XP_014998956.2
2327
91
XM_015143471.2
XP_014998957.2
2326
91
XM_015143475.2
XP_014998961.2
2326
91
XM_015143479.2
XP_014998965.2
2326
91
XM_015143487.2
XP_014998973.2
2310
91
XM_015143488.2
XP_014998974.2
1702
91
XM_028832010.1
XP_028687843.1
1772
91
XM_028832032.1
XP_028687865.1
2321
91
XM_028832044.1
XP_028687877.1
1702
91
XM_028832093.1
XP_028687926.1
2307
91
XM_028832123.1
XP_028687956.1
2328
91
XM_028832124.1
XP_028687957.1
2325
91
XM_028832125.1
XP_028687958.1
2323
91
XM_028832129.1
XP_028687962.1
2328
91
XM_028832131.1
XP_028687964.1
2311
91
XM_028832132.1
XP_028687965.1
2310
91
XM_028832133.1
XP_028687966.1
2328
91
XM_028832134.1
XP_028687967.1
1689
91
GenBank mRNA
JU333621.1
-
1591
91
JV043500.1
-
1557
91
JV576104.1
-
1608
91
JV710532.1
-
1620
91
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
G-protein coupled receptor
Macaca mulatta
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline